Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180572.2 | NP_850903.1 | 3 - 4 | 733 | 99 | |
GenBank mRNA | AF370232.1 | - | 3 - 4 | 695 | 99 | |
AY059914.1 | - | 3 - 4 | 715 | 99 | ||
AY088230.1 | - | 3 - 4 | 734 | 99 | ||
AY142540.1 | - | 3 - 4 | 658 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |