Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLA03 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036007.2 | NP_001031084.1 | 5 - 6 | 1633 | 138 | |
NM_001332604.1 | NP_001320605.1 | 5 - 6 | 1868 | 138 | ||
NM_102197.5 | NP_173762.4 | 6 - 7 | 1758 | 138 | ||
NM_180621.3 | NP_850952.1 | 7 - 8 | 1660 | 138 | ||
GenBank mRNA | AY099769.1 | - | 6 - 7 | 1413 | 138 | |
BT000203.1 | - | 6 - 7 | 1313 | 138 | ||
KJ138989.1 | - | 6 - 7 | 1313 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |