Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102272.4 | NP_564214.2 | 5 - 6 | 719 | 100 | |
NM_180622.3 | NP_850953.1 | 5 - 6 | 741 | 100 | ||
GenBank mRNA | AF015552.1 | - | 5 - 6 | 489 | 100 | |
AY088272.1 | - | 5 - 6 | 510 | 100 | ||
BT033157.1 | - | 5 - 6 | 483 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |