Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102272.4 | NP_564214.2 | 6 - 7 | 758 | 72 | |
NM_180622.3 | NP_850953.1 | 6 - 7 | 780 | 72 | ||
GenBank mRNA | AF015552.1 | - | 6 - 7 | 528 | 72 | |
AY088272.1 | - | 6 - 7 | 549 | 72 | ||
BT033157.1 | - | 6 - 7 | 522 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |