Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATPI4K ALPHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333375.1 | NP_001323160.1 | 13 - 14 | 4155 | 85 | |
NM_001333376.1 | NP_001323161.1 | 13 - 14 | 4141 | 85 | ||
NM_103824.4 | NP_175359.2 | 13 - 14 | 4174 | 85 | ||
NM_180629.2 | NP_850960.1 | 13 - 14 | 4176 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |