Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120500.3 | NP_196038.1 | 2 - 3 | 440 | 81 | |
GenBank mRNA | AK221569.1 | - | 2 - 3 | 440 | 81 | |
AY088872.1 | - | 2 - 3 | 436 | 81 | ||
DQ446916.1 | - | 2 - 3 | 393 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |