Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342755.1 | NP_001331162.1 | 8 - 9 | 1477 | 84 | |
NM_120504.2 | NP_568135.1 | 8 - 9 | 1361 | 84 | ||
NM_203000.2 | NP_974729.1 | 8 - 9 | 1335 | 84 | ||
GenBank mRNA | AB102952.1 | - | 8 - 9 | 1011 | 84 | |
FM213367.1 | - | 8 - 9 | 1167 | 84 | ||
FM213368.1 | - | 8 - 9 | 1070 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |