Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342767.1 | NP_001330089.1 | 27 - 28 | 2891 | 94 | |
NM_120518.5 | NP_196056.2 | 27 - 28 | 2904 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |