Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120523.4 | NP_196061.1 | 5 - 6 | 1618 | 104 | |
GenBank mRNA | AY099609.1 | - | 5 - 6 | 1491 | 104 | |
AY128859.1 | - | 5 - 6 | 1322 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |