Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342778.1 | NP_001330248.1 | 4 - 5 | 825 | 102 | |
NM_120531.3 | NP_196069.1 | 4 - 5 | 900 | 102 | ||
GenBank mRNA | BT021123.1 | - | 4 - 5 | 628 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |