Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342782.1 | NP_001332170.1 | 5 - 6 | 942 | 141 | |
NM_120544.3 | NP_196082.2 | 4 - 5 | 1175 | 141 | ||
NM_203002.4 | NP_974731.1 | 5 - 6 | 964 | 141 | ||
GenBank mRNA | AK119095.1 | - | 4 - 5 | 1175 | 141 | |
BT026507.1 | - | 3 - 4 | 366 | 141 | ||
DQ017966.1 | - | 5 - 6 | 783 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |