Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OFP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120564.2 | NP_196102.1 | 1 | 1290 | 80 | |
GenBank mRNA | AY084744.1 | - | 1 | 1066 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |