Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120575.3 | NP_568146.1 | 4 - 5 | 3447 | 76 | |
GenBank mRNA | AF175769.1 | - | 4 - 5 | 2651 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |