Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120611.3 | NP_196148.1 | 1 - 2 | 204 | 74 | |
GenBank mRNA | AY064035.1 | - | 1 - 2 | 204 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |