Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125703.2 | NP_001119175.1 | 2 - 3 | 355 | 142 | |
NM_120636.4 | NP_196173.1 | 2 - 3 | 355 | 142 | ||
GenBank mRNA | AK118437.1 | - | 2 - 3 | 331 | 142 | |
BT009685.1 | - | 2 - 3 | 218 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |