Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203303.1 | NP_001190232.1 | 7 - 8 | 869 | 68 | |
NM_120656.3 | NP_196193.1 | 6 - 7 | 867 | 68 | ||
NM_203007.3 | NP_974736.1 | 7 - 8 | 916 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |