Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342839.1 | NP_001318486.1 | 4 - 5 | 993 | 65 | |
NM_001342840.1 | NP_001330195.1 | 5 - 6 | 838 | 65 | ||
NM_120674.4 | NP_196211.1 | 5 - 6 | 943 | 65 | ||
GenBank mRNA | AY087829.1 | - | 5 - 6 | 902 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |