Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120698.5 | NP_196234.3 | 3 - 4 | 531 | 112 | |
GenBank mRNA | AY039944.1 | - | 3 - 4 | 472 | 112 | |
AY113862.1 | - | 3 - 4 | 396 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |