Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AtPOT1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120714.4 | NP_196249.2 | 2 - 3 | 379 | 79 | |
GenBank mRNA | AK221379.1 | - | 2 - 3 | 331 | 79 | |
AY884594.1 | - | 2 - 3 | 254 | 79 | ||
BT012568.1 | - | 2 - 3 | 254 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |