Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDX27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120717.5 | NP_196252.1 | 1 - 2 | 351 | 80 | |
GenBank mRNA | AF372941.1 | - | 1 - 2 | 284 | 80 | |
AY113163.1 | - | 1 - 2 | 223 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |