Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120765.3 | NP_196300.1 | 11 - 12 | 1594 | 130 | |
GenBank mRNA | AY518287.1 | - | 11 - 12 | 1441 | 130 | |
FJ708770.1 | - | 11 - 12 | 1316 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |