Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120783.4 | NP_568177.1 | 1 | 1595 | 144 | |
GenBank mRNA | AY099809.1 | - | 1 | 1253 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |