Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120811.5 | NP_196346.1 | 8 - 9 | 1918 | 134 | |
GenBank mRNA | AK226960.1 | - | 8 - 9 | 1662 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |