Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120812.4 | NP_568180.1 | 8 - 9 | 1103 | 87 | |
GenBank mRNA | AF389301.1 | - | 8 - 9 | 867 | 87 | |
BT010130.1 | - | 8 - 9 | 832 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |