Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120828.4 | NP_196363.1 | 1 - 2 | 749 | 95 | |
GenBank mRNA | BT010708.1 | - | 1 - 2 | 484 | 95 | |
BT012426.1 | - | 1 - 2 | 406 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |