Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120865.5 | NP_196400.2 | 3 - 4 | 664 | 67 | |
GenBank mRNA | BT015345.1 | - | 3 - 4 | 445 | 67 | |
BT015800.1 | - | 2 - 3 | 265 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |