Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPA70B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120884.2 | NP_196419.1 | 5 - 6 | 1028 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |