Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125718.1 | NP_001119190.1 | 10 - 11 | 2058 | 79 | |
NM_001203328.2 | NP_001190257.1 | 11 - 12 | 2005 | 79 | ||
NM_001203329.1 | NP_001190258.1 | 11 - 12 | 2005 | 79 | ||
NM_001203330.2 | NP_001190259.1 | 11 - 12 | 2014 | 79 | ||
NM_001342983.1 | NP_001318507.1 | 11 - 12 | 1841 | 79 | ||
NM_001342984.1 | NP_001318508.1 | 11 - 12 | 1838 | 79 | ||
NM_001342985.1 | NP_001330723.1 | 11 - 12 | 2002 | 79 | ||
NM_001342986.1 | NP_001330724.1 | 11 - 12 | 2017 | 79 | ||
GenBank mRNA | AB493742.1 | - | 8 - 9 | 1146 | 79 | |
BT029780.1 | - | 8 - 9 | 1146 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |