Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120909.4 | NP_196443.1 | 6 - 7 | 1273 | 77 | |
GenBank mRNA | AF370244.1 | - | 6 - 7 | 1159 | 77 | |
AY062969.1 | - | 6 - 7 | 1091 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |