Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335740.1 | NP_001324767.1 | 8 - 9 | 665 | 94 | |
NM_127700.4 | NP_565505.1 | 8 - 9 | 596 | 94 | ||
GenBank mRNA | AF324701.2 | - | 8 - 9 | 583 | 94 | |
AF326895.1 | - | 8 - 9 | 583 | 94 | ||
AF339713.1 | - | 8 - 9 | 568 | 94 | ||
AJ516948.1 | - | 8 - 9 | 568 | 94 | ||
BT000757.1 | - | 8 - 9 | 583 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |