Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202641.1 | NP_001189570.1 | 2 - 3 | 307 | 98 | |
NM_127719.4 | NP_179742.2 | 2 - 3 | 291 | 98 | ||
NM_201777.1 | NP_973506.1 | 2 - 3 | 282 | 98 | ||
GenBank mRNA | AK117731.1 | - | 2 - 3 | 282 | 98 | |
BT044606.1 | - | 2 - 3 | 124 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |