Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036312.2 | NP_001031389.1 | 4 - 5 | 965 | 61 | |
NM_001202642.2 | NP_001189571.1 | 4 - 5 | 965 | 61 | ||
NM_001335761.1 | NP_001318263.1 | 4 - 5 | 832 | 61 | ||
NM_001335762.1 | NP_001323476.1 | 4 - 5 | 846 | 61 | ||
NM_001335763.1 | NP_001323475.1 | 4 - 5 | 761 | 61 | ||
NM_127726.2 | NP_565514.1 | 4 - 5 | 846 | 61 | ||
GenBank mRNA | AK176420.1 | - | 4 - 5 | 812 | 61 | |
AY057587.1 | - | 4 - 5 | 848 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |