Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127748.4 | NP_179770.1 | 12 - 13 | 2012 | 84 | |
GenBank mRNA | AY035080.1 | - | 12 - 13 | 1918 | 84 | |
BT008573.1 | - | 12 - 13 | 1744 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |