Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036317.1 | NP_001031394.1 | 10 - 11 | 1395 | 71 | |
NM_127791.3 | NP_565529.1 | 9 - 10 | 1898 | 71 | ||
NM_179691.4 | NP_850022.1 | 10 - 11 | 1331 | 71 | ||
GenBank mRNA | AY064152.1 | - | 9 - 10 | 1380 | 71 | |
AY124811.1 | - | 9 - 10 | 1287 | 71 | ||
HM638413.1 | - | 9 - 10 | 1287 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |