Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127823.3 | NP_565539.1 | 4 - 5 | 744 | 74 | |
NM_201782.3 | NP_973511.1 | 3 - 4 | 842 | 74 | ||
GenBank mRNA | BT002359.1 | - | 4 - 5 | 658 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |