Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127845.4 | NP_179865.1 | 1 - 2 | 583 | 70 | |
GenBank mRNA | AK229218.1 | - | 1 - 2 | 500 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |