Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127866.3 | NP_179884.1 | 8 - 9 | 882 | 133 | |
NM_201788.2 | NP_973517.1 | 8 - 9 | 828 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |