Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335851.1 | NP_001323701.1 | 12 - 13 | 1592 | 116 | |
NM_127906.6 | NP_179923.2 | 12 - 13 | 1593 | 116 | ||
GenBank mRNA | BT001932.1 | - | 12 - 13 | 1377 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |