Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sks16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335859.1 | NP_001324736.1 | 3 - 4 | 644 | 74 | |
NM_127927.3 | NP_565554.1 | 3 - 4 | 540 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |