Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127962.3 | NP_179978.2 | 2 - 3 | 412 | 99 | |
GenBank mRNA | DQ446558.1 | - | 2 - 3 | 358 | 99 | |
DQ785742.1 | - | 2 - 3 | 358 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |