Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HHP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335921.1 | NP_001325271.1 | 2 - 3 | 518 | 96 | |
NM_127976.5 | NP_565564.1 | 3 - 4 | 633 | 96 | ||
GenBank mRNA | AF370179.1 | - | 3 - 4 | 412 | 96 | |
BT001975.1 | - | 3 - 4 | 371 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |