Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127982.4 | NP_179998.1 | 1 - 2 | 251 | 134 | |
GenBank mRNA | BT033153.1 | - | 1 - 2 | 207 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |