Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198510.1 | NP_001185439.1 | 4 - 5 | 465 | 65 | |
NM_106614.6 | NP_565216.2 | 4 - 5 | 525 | 65 | ||
NM_180651.3 | NP_850982.1 | 4 - 5 | 543 | 65 | ||
NM_202452.2 | NP_974181.1 | 4 - 5 | 455 | 65 | ||
GenBank mRNA | AY034912.1 | - | 4 - 5 | 434 | 65 | |
AY063103.1 | - | 4 - 5 | 298 | 65 | ||
AY088037.1 | - | 4 - 5 | 455 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |