Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126703.4 | NP_178744.3 | 2 - 3 | 170 | 73 | |
NM_180659.3 | NP_850990.1 | 2 - 3 | 206 | 73 | ||
GenBank mRNA | AY048212.1 | - | 2 - 3 | 119 | 73 | |
AY087310.1 | - | 2 - 3 | 170 | 73 | ||
AY094019.1 | - | 2 - 3 | 85 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |