Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtLPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112601.3 | NP_566570.3 | 1 - 2 | 360 | 76 | |
NM_180674.4 | NP_851005.1 | 1 - 2 | 443 | 76 | ||
GenBank mRNA | AF228640.1 | - | 1 - 2 | 357 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |