Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036800.2 | NP_001031877.1 | 10 - 11 | 1007 | 121 | |
NM_001343292.1 | NP_001318556.1 | 10 - 11 | 1007 | 121 | ||
NM_121378.5 | NP_568290.3 | 8 - 9 | 798 | 121 | ||
NM_180705.3 | NP_851036.1 | 10 - 11 | 1403 | 121 | ||
GenBank mRNA | AY039542.1 | - | 10 - 11 | 952 | 121 | |
AY062867.1 | - | 10 - 11 | 997 | 121 | ||
AY102150.1 | - | 10 - 11 | 889 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |