Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036800.2 | NP_001031877.1 | 2 - 3 | 299 | 74 | |
NM_001343292.1 | NP_001318556.1 | 2 - 3 | 299 | 74 | ||
NM_180705.3 | NP_851036.1 | 2 - 3 | 695 | 74 | ||
GenBank mRNA | AY039542.1 | - | 2 - 3 | 244 | 74 | |
AY062867.1 | - | 2 - 3 | 289 | 74 | ||
AY102150.1 | - | 2 - 3 | 181 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |