Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036844.3 | NP_001031921.3 | 9 - 10 | 995 | 87 | |
NM_122128.3 | NP_568413.1 | 9 - 10 | 995 | 87 | ||
GenBank mRNA | AF242580.1 | - | 9 - 10 | 1012 | 87 | |
AY136479.1 | - | 9 - 10 | 1069 | 87 | ||
BT006610.1 | - | 9 - 10 | 781 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |