Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alpha-ADR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343762.1 | NP_001330970.1 | 27 - 28 | 3343 | 106 | |
NM_180727.2 | NP_851058.1 | 28 - 29 | 3177 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |