Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122501.3 | NP_197972.2 | 10 - 11 | 1296 | 83 | |
NM_180746.3 | NP_851077.1 | 10 - 11 | 1296 | 83 | ||
GenBank mRNA | AY045681.1 | - | 10 - 11 | 1290 | 83 | |
AY090382.1 | - | 10 - 11 | 1259 | 83 | ||
L11454.1 | - | 10 - 11 | 1292 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |